Assay Details
| Gene Symbol: B130011K05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK044916.1 | - | 1 | 91 | ||
| AK050681.1 | - | 1 | 727 | 91 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

B130011K05Rik| Gene Symbol: B130011K05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK044916.1 | - | 1 | 91 | ||
| AK050681.1 | - | 1 | 727 | 91 | ||
Set Membership: |
|