Assay Details
Gene Symbol: Gm10535 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK134881.1 | - | 1 - 2 | 89 | 89 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK134881.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
