Assay Details
| Gene Symbol: Mgrn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252437.1 | NP_001239366.1 | 17 | 2298 | 113 | |
| NM_029657.4 | NP_083933.1 | 17 | 2295 | 113 | ||
| GenBank mRNA | AK088533.1 | - | 17 | 2078 | 113 | |
| AK129161.1 | - | 13 | 1661 | 113 | ||
| AK150176.1 | - | 18 | 2131 | 113 | ||
| BC046830.1 | - | 17 | 2144 | 6811606 | 113 | |
More Information
Set Membership: |
|

Mgrn1
Genomic Map