Assay Details
Gene Symbol: Gm5486 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK135986.1 | - | 2 - 3 | 330 | 58 | |
AK139943.1 | - | 357 | 58 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK139943.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
