Assay Details
| Gene Symbol: Atp5o | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138597.2 | NP_613063.1 | 4 | 393 | 93 | |
| GenBank mRNA | AK005309.1 | - | 4 | 392 | 93 | |
| AK168292.1 | - | 4 | 348 | 93 | ||
| AK169318.1 | - | 4 | 327 | 93 | ||
| BC012241.1 | - | 4 | 341 | 4023318 | 93 | |
| BC096643.1 | - | 4 | 351 | 93 | ||
More Information
Set Membership: |
|

Atp5o
Genomic Map