Assay Details
| Gene Symbol: Sec61b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024171.2 | NP_077133.1 | 2 - 3 | 186 | 78 | |
| GenBank mRNA | AK004505.1 | - | 2 - 3 | 187 | 78 | |
| AK008468.2 | - | 629 | 78 | |||
| AK008739.1 | - | 2 - 3 | 190 | 78 | ||
| BC002089.1 | - | 2 - 3 | 158 | 3487410 | 78 | |
| BC081445.1 | - | 2 - 3 | 154 | 6412192 | 78 | |
More Information
Set Membership: |
|

Sec61b
Genomic Map