Assay Details
| Gene Symbol: Vdac3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198998.1 | NP_001185927.1 | 10 | 1119 | 129 | |
| NM_011696.2 | NP_035826.1 | 9 | 1116 | 129 | ||
| GenBank mRNA | AK002296.1 | - | 9 | 1063 | 129 | |
| AK083785.1 | - | 10 | 1047 | 129 | ||
| AK146957.1 | - | 10 | 1072 | 129 | ||
| AK148744.1 | - | 3 | 360 | 129 | ||
| AK150741.1 | - | 9 | 1058 | 129 | ||
| AK150828.1 | - | 10 | 1047 | 129 | ||
| AK159431.1 | - | 9 | 1058 | 129 | ||
| AK161283.1 | - | 9 | 1045 | 129 | ||
| AK167781.1 | - | 9 | 1061 | 129 | ||
| AK168815.1 | - | 10 | 1064 | 129 | ||
| AK169099.1 | - | 10 | 1072 | 129 | ||
| AK169228.1 | - | 9 | 1044 | 129 | ||
| AK199811.1 | - | 2 | 243 | 128 | ||
| BC004743.1 | - | 9 | 1014 | 3583800 | 129 | |
| BC089336.1 | - | 10 | 1036 | 30357677 | 129 | |
| U30839.1 | - | 9 | 1061 | 129 | ||
More Information
Set Membership: |
|

Vdac3
Genomic Map