Assay Details
| Gene Symbol: Agk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023538.2 | NP_076027.1 | 8 - 9 | 567 | 95 | |
| GenBank mRNA | AJ401619.1 | - | 8 - 9 | 540 | 95 | |
| AK076224.1 | - | 7 - 8 | 530 | 95 | ||
| AY772469.1 | - | 7 - 8 | 508 | 95 | ||
| BC019145.1 | - | 8 - 9 | 561 | 5051535 | 95 | |
| BC093525.1 | - | 8 - 9 | 537 | 5151866 | 95 | |
More Information
Set Membership: |
|

Agk
Genomic Map