Assay Details
| Gene Symbol: Agk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023538.2 | NP_076027.1 | 9 - 10 | 644 | 70 | |
| GenBank mRNA | AJ401619.1 | - | 9 - 10 | 617 | 70 | |
| AK076224.1 | - | 8 - 9 | 607 | 70 | ||
| AY772469.1 | - | 8 - 9 | 585 | 70 | ||
| BC019145.1 | - | 9 - 10 | 638 | 5051535 | 70 | |
| BC093525.1 | - | 9 - 10 | 614 | 5151866 | 70 | |
More Information
Set Membership: |
|

Agk
Genomic Map