Assay Details
| Gene Symbol: Rbmxl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252089.1 | NP_001239018.1 | 2 | 1260 | 101 | |
| NM_009033.2 | NP_033059.2 | 2 | 1306 | 101 | ||
| GenBank mRNA | AF031568.1 | - | 2 | 1172 | 101 | |
| AK041927.1 | - | 2 | 1197 | 101 | ||
| AK088213.1 | - | 2 | 1189 | 100 | ||
| AK149658.1 | - | 2 | 1279 | 101 | ||
| AK149764.1 | - | 2 | 1279 | 101 | ||
| BC011441.1 | - | 2 | 1211 | 3153831 | 101 | |
| BC089350.1 | - | 2 | 1220 | 30359767 | 101 | |
More Information
Set Membership: |
|

Rbmxl1
Genomic Map