Assay Details
Gene Symbol: Nrm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK143140.1 | - | 1926 | 70 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Nrm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK143140.1 | - | 1926 | 70 |