| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix HT MG-430 PM Array Plate |
1428083_PM_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1428083_PM_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1428083_at |
- |
- |
| Affymetrix Mouse Clariom S Array |
PSR1900008982.mm.1 |
NR_003513.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900008982.mm.1 |
TC1900000986.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900008969.mm.1 |
NR_003513.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900008969.mm.1 |
TC1900000986.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900008976.mm.1 |
NR_003513.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900008976.mm.1 |
TC1900000986.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900008986.mm.1 |
NR_003513.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900008986.mm.1 |
TC1900000986.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900008971.mm.1 |
NR_003513.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900008971.mm.1 |
TC1900000986.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900008970.mm.1 |
NR_003513.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900008970.mm.1 |
TC1900000986.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900008980.mm.1 |
NR_003513.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900008980.mm.1 |
TC1900000986.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900008983.mm.1 |
NR_003513.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900008983.mm.1 |
TC1900000986.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900008985.mm.1 |
NR_003513.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900008985.mm.1 |
TC1900000986.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900008972.mm.1 |
NR_003513.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900008972.mm.1 |
TC1900000986.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900008975.mm.1 |
NR_003513.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900008975.mm.1 |
TC1900000986.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900008978.mm.1 |
NR_003513.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900008978.mm.1 |
TC1900000986.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900008973.mm.1 |
NR_003513.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900008973.mm.1 |
TC1900000986.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900008984.mm.1 |
NR_003513.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900008984.mm.1 |
TC1900000986.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1900008968.mm.1 |
NR_003513.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1900008968.mm.1 |
TC1900000986.mm.1 |
MTA TC |
| Mouse Expression Set 430 |
1428083_PM_at |
- |
- |
| Mouse Expression Set 430 |
1428083_at |
- |
- |
| Mouse Genome 430A 2.0 Array |
1428083_PM_at |
- |
- |
| Mouse Genome 430A 2.0 Array |
1428083_at |
- |
- |
| Affymetrix HT Mouse Genome 430A Array |
1428083_PM_at |
- |
- |
| Affymetrix HT Mouse Genome 430A Array |
1428083_at |
- |
- |