Assay Details
Gene Symbol: BE692007 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK140345.1 | - | 1098 | 76 |
Gene Symbol: Ms4a6c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK143011.1 | - | 1 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
