Assay Details
| Gene Symbol: Ldhb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK002257.1 | - | 7 | 893 | 73 | |
| AK019391.1 | - | 7 | 930 | 73 | ||
| AK131637.1 | - | 7 | 918 | 73 | ||
| BC046755.1 | - | 7 | 915 | 5693226 | 73 | |
| BC087888.1 | - | 4 | 312 | 73 | ||
| BC148214.1 | - | 3 | 367 | 73 | ||
| X51905.1 | - | 7 | 864 | 73 | ||
More Information
Set Membership: |
|

Ldhb
Genomic Map