Assay Details
| Gene Symbol: Gm14342 | |||||||
|---|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | DQ860414.1 | - | LIT3342 HAR1R |
1 | 711 | 101 | |
| DQ860415.1 | - | LIT3343 HAR1R |
815 | 101 | |||
More Information
Set Membership: |
|

Gm14342
Genomic Map