Assay Details
| Gene Symbol: Dnm3os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_002870.2 | 1 | 3884 | 99 | ||
| GenBank mRNA | AK053654.1 | - | 1 | 2668 | 99 | |
| AK164873.1 | - | 1 | 1712 | 99 | ||
| AY751519.1 | - | 1 | 3844 | 99 | ||
More Information
Set Membership: |
|

Dnm3os
Genomic Map