Assay Details
| Gene Symbol: Cat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009804.2 | NP_033934.2 | 2 | 243 | 82 | |
| GenBank mRNA | AK075853.1 | - | 2 | 213 | 82 | |
| AK088168.1 | - | 2 | 205 | 82 | ||
| AK128919.1 | - | 2 | 218 | 82 | ||
| AK133881.1 | - | 2 | 211 | 82 | ||
| AK148961.1 | - | 2 | 214 | 79 | ||
| AK150893.1 | - | 2 | 214 | 82 | ||
| AK159152.1 | - | 2 | 214 | 82 | ||
| AK159885.1 | - | 2 | 214 | 82 | ||
| AK159891.1 | - | 2 | 165 | 82 | ||
| AK159913.1 | - | 2 | 195 | 82 | ||
| AK169069.1 | - | 2 | 208 | 82 | ||
| AY040626.1 | - | 2 | 184 | 82 | ||
| BC013447.1 | - | 2 | 200 | 4241333 | 82 | |
| BC047126.1 | - | 2 | 158 | 82 | ||
| BY093963.1 | - | 2 | 243 | 82 | ||
| L25069.1 | - | 2 | 215 | 82 | ||
| M62897.1 | - | 2 | 128 | 82 | ||
| X52108.1 | - | 2 | 128 | 82 | ||
More Information
Set Membership: |
|

Cat
Genomic Map