Assay Details
Gene Symbol: Tmx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029148.1 | NP_083424.1 | 8 | 2903 | 107 | |
GenBank mRNA | AK013100.1 | - | 1 | 87 | 107 | |
AK129301.1 | - | 8 | 2902 | 107 | ||
AK147465.1 | - | 8 | 2904 | 107 | ||
BC050918.1 | - | 8 | 2802 | 107 | ||
BC054558.1 | - | 8 | 2903 | 6405599 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
