Assay Details
Gene Symbol: A630075K04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK042254.1 | - | 1949 | 66 |
Gene Symbol: AU022855 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK080354.1 | - | 1 | 1235 | 66 |
Gene Symbol: Sgip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK043905.1 | - | 803 | 66 | ||
AK090117.1 | - | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
