Assay Details
| Gene Symbol: Ifi203 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF022371.1 | - | 1757 | 94 | ||
| AK144663.1 | - | 9 | 1822 | 94 | ||
| AK164695.1 | - | 6 | 1534 | 94 | ||
| L14559.1 | - | 5 | 1333 | 94 | ||
More Information
Set Membership: |
|

Ifi203
Genomic Map