Assay Details
| Gene Symbol: Tox4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023434.3 | NP_075923.2 | 10 | 2504 | 68 | |
| GenBank mRNA | AF228408.1 | - | 8 | 2163 | 68 | |
| AK019980.1 | - | 6 | 1692 | 68 | ||
| AK088144.1 | - | 9 | 2177 | 68 | ||
| AK137050.1 | - | 728 | 68 | |||
| AK147023.1 | - | 9 | 2210 | 68 | ||
| AK147834.1 | - | 9 | 2181 | 68 | ||
| AK153278.1 | - | 9 | 2170 | 68 | ||
| AK173012.1 | - | 9 | 2208 | 68 | ||
| BC020135.1 | - | 1 | 116 | 68 | ||
| BC050091.1 | - | 8 | 2160 | 30023547 | 68 | |
More Information
Set Membership: |
|

Tox4
Genomic Map