Assay Details
| Gene Symbol: Nmnat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133435.1 | NP_597679.1 | 2 | 118 | 90 | |
| GenBank mRNA | AF260925.1 | - | 2 | 271 | 90 | |
| AK011137.1 | - | 2 | 140 | 90 | ||
| AK012194.1 | - | 2 | 352 | 90 | ||
| AK017631.1 | - | 128 | 90 | |||
| AK152407.1 | - | 178 | 90 | |||
| AY679721.1 | - | 2 | 118 | 90 | ||
| BC138500.1 | - | 2 | 186 | 8861520 | 90 | |
| BC138502.1 | - | 2 | 186 | 8861522 | 90 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK011137.1, AK012194.1, AY679721.1, BC138500.1, BC138502.1, NM_133435.1 transcripts.
Set Membership: |
|

Nmnat1
Genomic Map