Assay Details
| Gene Symbol: Mul1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026689.3 | NP_080965.2 | 4 | 1850 | 69 | |
| GenBank mRNA | AK076419.1 | - | 4 | 1811 | 69 | |
| AK083295.1 | - | 4 | 1822 | 69 | ||
| AK153956.1 | - | 4 | 1800 | 69 | ||
More Information
Set Membership: |
|

Mul1
Genomic Map