Assay Details
| Gene Symbol: Rnf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF169300.1 | - | 8 | 2510 | 95 | |
| AK019171.1 | - | 8 | 2525 | 95 | ||
| AK019656.1 | - | 8 | 2563 | 95 | ||
| AK037465.1 | - | 1927 | 95 | |||
| AK090162.1 | - | 8 | 2551 | 95 | ||
| AK145596.1 | - | 8 | 2489 | 95 | ||
| AK147057.1 | - | 8 | 2472 | 95 | ||
| AK151116.1 | - | 8 | 2520 | 95 | ||
| AK158987.1 | - | 8 | 2529 | 95 | ||
| AK159296.1 | - | 8 | 2527 | 95 | ||
| AK159778.1 | - | 8 | 2521 | 95 | ||
| AK159949.1 | - | 8 | 2569 | 95 | ||
| BC003282.1 | - | 8 | 2491 | 3582133 | 95 | |
| BC005551.1 | - | 1 | 1477 | 95 | ||
More Information
Set Membership: |
|

Rnf4
Genomic Map