Assay Details
| Gene Symbol: Zc3hc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172735.2 | NP_766323.1 | 11 | 1732 | 73 | |
| GenBank mRNA | AK004251.1 | - | 1 | 716 | 73 | |
| AK088527.1 | - | 11 | 1733 | 73 | ||
| AK146228.1 | - | 10 | 1703 | 73 | ||
| AK151694.1 | - | 10 | 1684 | 73 | ||
| AK167465.1 | - | 10 | 1689 | 73 | ||
| AK167992.1 | - | 10 | 1703 | 73 | ||
| AK168071.1 | - | 10 | 1698 | 73 | ||
| AK209395.1 | - | 1 | 168 | 73 | ||
| BC024560.1 | - | 4 | 704 | 73 | ||
| BC037445.1 | - | 10 | 1642 | 73 | ||
| BC050141.1 | - | 10 | 1681 | 30011212 | 73 | |
More Information
Set Membership: |
|

Zc3hc1
Genomic Map