Assay Details
| Gene Symbol: Rn4.5s | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_002841.1 | 103 | 73 | |||
| GenBank mRNA | AK040427.1 | - | 103 | 73 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rn4.5s| Gene Symbol: Rn4.5s | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_002841.1 | 103 | 73 | |||
| GenBank mRNA | AK040427.1 | - | 103 | 73 | ||
Set Membership: |
|