Assay Details
| Gene Symbol: B3gnt8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036740.2 | NP_001031817.1 | 2 | 530 | 101 | |
| NM_146184.4 | NP_666296.1 | 1 | 922 | 101 | ||
| GenBank mRNA | AK042388.1 | - | 1 | 905 | 101 | |
| AK154220.1 | - | 517 | 101 | |||
| AK154901.1 | - | 2 | 486 | 101 | ||
| BC025206.1 | - | 2 | 516 | 5037295 | 101 | |
More Information
Set Membership: |
|

B3gnt8
Genomic Map