Assay Details
Gene Symbol: Snord116 | |||||||
---|---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF241256.1 | - | LIT1355 PWCR1 |
2122 | 63 |
Gene Symbol: Ipw | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK045353.1 | - | 1 | 1074 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
