Assay Details
| Gene Symbol: Hnrnpk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301340.1 | NP_001288269.1 | 4 - 5 | 354 | 69 | |
| NM_001301341.1 | NP_001288270.1 | 6 - 7 | 481 | 69 | ||
| NM_001301343.1 | NP_001288272.1 | 4 - 5 | 340 | 69 | ||
| NM_001301344.1 | NP_001288273.1 | 4 - 5 | 354 | 69 | ||
| NM_001301345.1 | NP_001288274.1 | 4 - 5 | 354 | 69 | ||
| NM_025279.3 | NP_079555.1 | 4 - 5 | 354 | 69 | ||
| GenBank mRNA | AK011428.1 | - | 4 - 5 | 359 | 69 | |
| AK028023.1 | - | 4 - 5 | 288 | 69 | ||
| AK051313.1 | - | 4 - 5 | 348 | 69 | ||
| AK078777.1 | - | 6 - 7 | 482 | 69 | ||
| AK088462.1 | - | 4 - 5 | 341 | 69 | ||
| AK128891.1 | - | 4 - 5 | 347 | 69 | ||
| AK135519.1 | - | 4 - 5 | 268 | 69 | ||
| AK144245.1 | - | 7564 | 69 | |||
| AK150848.1 | - | 4 - 5 | 334 | 69 | ||
| AK151689.1 | - | 4 - 5 | 308 | 69 | ||
| AK152927.1 | - | 4 - 5 | 308 | 69 | ||
| AK160615.1 | - | 4 - 5 | 324 | 69 | ||
| AK160887.1 | - | 4 - 5 | 356 | 69 | ||
| AK166656.1 | - | 4 - 5 | 359 | 69 | ||
| AK166927.1 | - | 4 - 5 | 312 | 69 | ||
| AK167600.1 | - | 4 - 5 | 345 | 69 | ||
| AK169064.1 | - | 4 - 5 | 344 | 69 | ||
| BC006694.1 | - | 4 - 5 | 321 | 3583037 | 69 | |
| BC089328.1 | - | 4 - 5 | 277 | 6402614 | 69 | |
| EU604547.1 | - | 3 - 4 | 207 | 69 | ||
More Information
Set Membership: |
|

Hnrnpk
Genomic Map