Assay Details
| Gene Symbol: Atp11a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293667.1 | NP_001280596.1 | 30 | 7433 | 114 | |
| NM_001293668.1 | NP_001280597.1 | 30 | 7414 | 114 | ||
| NM_015804.3 | NP_056619.1 | 29 | 7329 | 114 | ||
| NR_121603.1 | 30 | 7534 | 114 | |||
| GenBank mRNA | AK016173.1 | - | 1 | 691 | 114 | |
| AK028779.1 | - | 1 | 2174 | 114 | ||
| AK147497.1 | - | 30 | 7436 | 114 | ||
| AK161916.1 | - | 1 | 2356 | 114 | ||
More Information
Set Membership: |
|

Atp11a
Genomic Map