Assay Details
| Gene Symbol: Pafah1b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008775.3 | NP_032801.2 | 6 | 4130 | 99 | |
| GenBank mRNA | AK053415.1 | - | 1 | 2426 | 99 | |
| AK169746.1 | - | 1 | 2427 | 99 | ||
More Information
Set Membership: |
|

Pafah1b2
Genomic Map