Assay Details
Gene Symbol: Sparc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290817.1 | NP_001277746.1 | 10 | 1470 | 146 | |
NM_009242.5 | NP_033268.1 | 10 | 1473 | 146 | ||
GenBank mRNA | AA798908.1 | - | 1 | 98 | 145 | |
AK014286.1 | - | 10 | 1276 | 146 | ||
AK133377.1 | - | 10 | 1244 | 146 | ||
AK146935.1 | - | 10 | 1245 | 146 | ||
AK148670.1 | - | 10 | 1256 | 146 | ||
AK161229.1 | - | 10 | 1260 | 146 | ||
X04017.1 | - | 10 | 1256 | 146 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
