Assay Details
| Gene Symbol: Kansl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081045.1 | NP_001074514.1 | 2 | 1254 | 125 | |
| GenBank mRNA | AK037800.1 | - | 3 | 820 | 125 | |
| AK122484.1 | - | 2 | 1260 | 125 | ||
| AK153679.1 | - | 2 | 1235 | 125 | ||
| AK161514.1 | - | 2 | 769 | 125 | ||
| BC043121.1 | - | 2 | 803 | 6409084 | 125 | |
| BC053389.1 | - | 2 | 804 | 30061629 | 125 | |
| BC054752.1 | - | 3 | 1077 | 6834541 | 125 | |
| BC079594.1 | - | 2 | 802 | 6827379 | 125 | |
More Information
Set Membership: |
|

Kansl1
Genomic Map