Assay Details
| Gene Symbol: Gm11772 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BB323723.2 | - | 1 | 322 | 107 | |
| BX526074.1 | - | 2 | 227 | 107 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm11772| Gene Symbol: Gm11772 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BB323723.2 | - | 1 | 322 | 107 | |
| BX526074.1 | - | 2 | 227 | 107 | ||
Set Membership: |
|