Assay Details
| Gene Symbol: H2afz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016750.3 | NP_058030.1 | 5 | 937 | 108 | |
| GenBank mRNA | AK088704.1 | - | 5 | 884 | 108 | |
| AK143572.1 | - | 5 | 836 | 108 | ||
| AK146181.1 | - | 5 | 869 | 108 | ||
| AK151052.1 | - | 5 | 836 | 108 | ||
| AK152848.1 | - | 5 | 835 | 108 | ||
| AK169003.1 | - | 5 | 722 | 108 | ||
| AK213534.1 | - | 1 | 227 | 108 | ||
| U70494.1 | - | 5 | 825 | 108 | ||
More Information
Set Membership: |
|

H2afz
Genomic Map