Assay Details
| Gene Symbol: Zfp213 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033496.3 | NP_001028668.2 | 6 | 2546 | 125 | |
| GenBank mRNA | AK133902.1 | - | 6 | 2446 | 125 | |
| AK147992.1 | - | 6 | 2450 | 125 | ||
| AK171979.1 | - | 2 | 1134 | 125 | ||
| BX528154.1 | - | 1 | 125 | |||
More Information
Set Membership: |
|

Zfp213
Genomic Map