Assay Details
| Gene Symbol: Cks2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025415.3 | NP_079691.1 | 2 | 320 | 86 | |
| GenBank mRNA | AK002451.1 | - | 2 | 228 | 86 | |
| AK004163.1 | - | 2 | 228 | 86 | ||
| AK077690.1 | - | 2 | 130 | 86 | ||
| AK166688.1 | - | 2 | 237 | 86 | ||
| BC022647.1 | - | 2 | 227 | 4216297 | 86 | |
| BC094919.1 | - | 2 | 196 | 6431137 | 86 | |
| DV661312.1 | - | 2 | 320 | 86 | ||
More Information
Set Membership: |
|

Cks2
Genomic Map