Assay Details
| Gene Symbol: Dleu2 | |||||||
|---|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF380425.1 | - | LIT3421 DLEU2 |
1 | 64 | 64 | |
| AK029956.1 | - | 1 | 90 | 64 | |||
More Information
Set Membership: |
|

Dleu2
Genomic Map