Assay Details
| Gene Symbol: Dhrs7b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172112.1 | NP_001165583.1 | 5 - 6 | 728 | 68 | |
| NM_145428.2 | NP_663403.1 | 5 - 6 | 699 | 68 | ||
| GenBank mRNA | AK052209.1 | - | 5 - 6 | 651 | 68 | |
| AK139084.1 | - | 5 - 6 | 680 | 68 | ||
| AK144057.1 | - | 5 - 6 | 634 | 68 | ||
| BC003479.1 | - | 5 - 6 | 654 | 2655855 | 68 | |
| JX196872.1 | - | 5 - 6 | 618 | 68 | ||
More Information
Set Membership: |
|

Dhrs7b
Genomic Map