Assay Details
Gene Symbol: Wdr53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001185162.1 | NP_001172091.1 | 1 - 2 | 39 | 70 | |
GenBank mRNA | BY142325.1 | - | 1 - 2 | 40 | 70 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001185162.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
