Assay Details
| Gene Symbol: Wdr53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001185162.1 | NP_001172091.1 | 2 - 3 | 539 | 108 | |
| NM_026898.2 | NP_081174.1 | 2 - 3 | 771 | 108 | ||
| GenBank mRNA | AK005105.1 | - | 2 - 3 | 738 | 108 | |
| BC028850.1 | - | 1 - 2 | 503 | 3987279 | 108 | |
More Information
Set Membership: |
|

Wdr53
Genomic Map