Assay Details
| Gene Symbol: Aldoart1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199270.1 | NP_001186199.1 | 1 | 1515 | 119 | |
| GenBank mRNA | AK014956.1 | - | 1 | 1516 | 119 | |
| AK016845.1 | - | 1 | 1488 | 119 | ||
More Information
Set Membership: |
|

Aldoart1
Genomic Map