Assay Details
| Gene Symbol: Clec10a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204252.1 | NP_001191181.1 | 2 - 3 | 378 | 85 | |
| GenBank mRNA | AK140762.1 | - | 1 - 2 | 1348 | 85 | |
| AK170779.1 | - | 2 - 3 | 335 | 85 | ||
More Information
Set Membership: |
|

Clec10a
Genomic Map