Assay Details
| Gene Symbol: Arid3c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017362.2 | NP_001017362.1 | 5 - 6 | 982 | 91 | |
| NM_001252622.1 | NP_001239551.1 | 5 - 6 | 982 | 91 | ||
| GenBank mRNA | BC141237.1 | - | 4 - 5 | 951 | 9055849 | 91 |
| BC145563.1 | - | 4 - 5 | 951 | 9054128 | 91 | |
More Information
Set Membership: |
|

Arid3c
Genomic Map