Assay Details
Gene Symbol: Smim22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001253796.1 | NP_001240725.1 | 3 - 4 | 101 | ||
NM_001253797.1 | NP_001240726.1 | 3 - 4 | 336 | 86 | ||
NM_001253802.1 | NP_001240731.1 | 3 - 4 | 101 | |||
NM_001253803.1 | NP_001240732.1 | 3 - 4 | 271 | 86 | ||
NR_045595.1 | 3 - 4 | 101 | ||||
NR_045596.1 | 2 - 3 | 101 | ||||
GenBank mRNA | AW985536.1 | - | 3 - 4 | 262 | 86 | |
BB610432.1 | - | 2 - 3 | 173 | 86 | ||
BC047216.1 | - | 3 - 4 | 101 | |||
CJ237462.1 | - | 3 - 4 | 333 | 86 | ||
CJ320043.1 | - | 3 - 4 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
