Assay Details
| Gene Symbol: Echdc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001254754.1 | NP_001241683.1 | 8 - 9 | 837 | 78 | |
| NM_026728.4 | NP_081004.2 | 9 - 10 | 905 | 78 | ||
| GenBank mRNA | AI839329.1 | - | 1 - 2 | 418 | 78 | |
| AK005030.1 | - | 9 - 10 | 846 | 78 | ||
| AK011365.1 | - | 8 - 9 | 783 | 78 | ||
| AK153878.1 | - | 9 - 10 | 880 | 78 | ||
| AK160482.1 | - | 7 - 8 | 608 | 78 | ||
| AK166388.1 | - | 9 - 10 | 877 | 78 | ||
| AK166965.1 | - | 9 - 10 | 847 | 78 | ||
| BC025104.1 | - | 9 - 10 | 851 | 3591648 | 78 | |
| BC025454.1 | - | 5 - 6 | 389 | 5352625 | 78 | |
More Information
Set Membership: |
|

Echdc2
Genomic Map