Assay Details
| Gene Symbol: Cass4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080820.2 | NP_001074289.1 | 3 - 4 | 734 | 103 | |
| NM_001276422.1 | NP_001263351.1 | 3 - 4 | 734 | 103 | ||
| NM_001276423.1 | NP_001263352.1 | 3 - 4 | 734 | 103 | ||
| GenBank mRNA | AK036419.1 | - | 3 - 4 | 708 | 103 | |
| AK080960.1 | - | 3 - 4 | 708 | 103 | ||
| AK089450.1 | - | 3 - 4 | 735 | 103 | ||
| AK170726.1 | - | 3 - 4 | 734 | 103 | ||
| BC117858.1 | - | 3 - 4 | 628 | 40095182 | 103 | |
| BC117859.1 | - | 3 - 4 | 627 | 40095183 | 103 | |
| BC129975.1 | - | 3 - 4 | 625 | 40095184 | 103 | |
| BC129976.1 | - | 3 - 4 | 627 | 40095185 | 103 | |
More Information
Set Membership: |
|

Cass4
Genomic Map