Assay Details
| Gene Symbol: Cass4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276422.1 | NP_001263351.1 | 4 - 5 | 818 | 107 | |
| GenBank mRNA | BC129976.1 | - | 4 - 5 | 711 | 40095185 | 107 |
More Information
Set Membership: |
|

Cass4
Genomic Map