Assay Details
| Gene Symbol: Prnp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278256.1 | NP_001265185.1 | 3 - 4 | 1718 | 66 | |
| GenBank mRNA | AK159056.1 | - | 3 - 4 | 1682 | 66 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK159056.1, NM_001278256.1 transcripts.
Set Membership: |
|

Prnp
Genomic Map