Assay Details
| Gene Symbol: Zfp386 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004066.3 | NP_001004066.1 | 1 - 2 | 147 | 104 | |
| NM_001286214.1 | NP_001273143.1 | 1 - 2 | 147 | 104 | ||
| NM_019565.4 | NP_062511.2 | 1 - 2 | 147 | 104 | ||
| GenBank mRNA | AF175222.1 | - | 1 - 2 | 95 | 104 | |
| AK043310.1 | - | 1 - 2 | 81 | 104 | ||
| AK165544.1 | - | 1 - 2 | 129 | 104 | ||
| AK220544.1 | - | 1 - 2 | 95 | 104 | ||
| BC004747.1 | - | 1 - 2 | 94 | 3584034 | 104 | |
| BE652367.1 | - | 1 - 2 | 95 | 104 | ||
| CF723896.1 | - | 1 - 2 | 93 | 104 | ||
| CJ140586.1 | - | 1 - 2 | 148 | 104 | ||
| CJ172693.1 | - | 1 - 2 | 113 | 104 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF175222.1, AK165544.1, BC004747.1, NM_001004066.3, NM_001286214.1, NM_019565.4 transcripts.
Set Membership: |
|

Zfp386
Genomic Map